Learn More
Sigma Aldrich Fine Chemicals Biosciences Magnesium sulfate heptahydrate, 10034-99-8, MFCD00149785, 500 G
Catalog No. NC0619479
Encompass
Linear Formula: MgSO4 · 7H2O, Molecular Weight: 246.47, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%, Synonym: Epsom salts. Magnesium sulfate heptahydrate has been used: in the preparation of Scott′s tap water substitute as a bluing reagent for nuclear staining, as a component of stabilization solution for the isolation of pituitary cells for matrix-assisted laser desorption/ionization mass spectroscopy (MALDI-MS), and as an additive in the preparation of artificial sea water for the maintenance of Aplysia californica and in modified Gey′s balanced salt solution.
Catalog No. NC0619479 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M2773500G
Linear Formula: MgSO4 · 7H2O, Molecular Weight: 246.47, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%, Synonym: Epsom salts. Magnesium sulfate heptahydrate has been used: in the preparation of Scott′s tap water substitute as a bluing reagent for nuclear staining, as a component of stabilization solution for the isolation of pituitary cells for matrix-assisted laser desorption/ionization mass spectroscopy (MALDI-MS), and as an additive in the preparation of artificial sea water for the maintenance of Aplysia californica and in modified Gey′s balanced salt solution.