Learn More
Alkali Scientific Magnesium Sulfate Hepta Hydrate, 500 G
SDP
Catalog No. 5000661663
Encompass_Preferred
Magnesium Sulfate Heptahydrate is a high-purity crystalline reagent used in various laboratory applications requiring magnesium ions. The 500g format provides an efficient source of magnesium for processes such as enzyme catalysis, microbial growth, and biochemical assays. Magnesium ions are essential for the activity of many enzymes, especially those involved in energy transfer, DNA replication, and protein synthesis. This reagent is commonly used in cell culture media, buffers, and enzyme reaction mixtures. It also finds applications in agriculture and horticulture, where magnesium is crucial for plant growth and photosynthesis, making it a versatile compound for scientific research and industrial use.
Catalog No. 50-006-61663 Supplier Alkali Scientific Supplier No. MAG002CP
Magnesium Sulfate Heptahydrate is a high-purity crystalline reagent used in various laboratory applications requiring magnesium ions. The 500g format provides an efficient source of magnesium for processes such as enzyme catalysis, microbial growth, and biochemical assays. Magnesium ions are essential for the activity of many enzymes, especially those involved in energy transfer, DNA replication, and protein synthesis. This reagent is commonly used in cell culture media, buffers, and enzyme reaction mixtures. It also finds applications in agriculture and horticulture, where magnesium is crucial for plant growth and photosynthesis, making it a versatile compound for scientific research and industrial use.