Learn More
Strem, An Ascensus Company CAS# 10034-99-8. 1kg. Magnesium sulfate heptahydrate, 98+% (ACS). MFCD00149785
SDP
Catalog No. 5090113964
Encompass
CAS# 10034-99-8. 1kg. Magnesium sulfate heptahydrate, 98+% (ACS). MFCD00149785. Molecular Weight: 120.39 (246.48). Molecular Formula: MgSO4∙7H2O. Color/form: white xtl. Strem# 93-1256. http://www.strem.com/catalog/v/93-1256/
Catalog No. 50-901-13964 Supplier Strem, An Ascensus Company Supplier No. 9312561000G
CAS# 10034-99-8. 1kg. Magnesium sulfate heptahydrate, 98+% (ACS). MFCD00149785. Molecular Weight: 120.39 (246.48). Molecular Formula: MgSO4∙7H2O. Color/form: white xtl. Strem# 93-1256. http://www.strem.com/catalog/v/93-1256/