Learn More
Alkali Scientific Magnesium Sulfate Heptahydrate, 100 G
SDP
Catalog No. 5000661483
Encompass_Preferred
Magnesium Sulfate Heptahydrate (MAG003 CP) is a laboratory-grade salt used in a variety of chemical and biochemical applications. This compound is a crystalline form of magnesium sulfate, commonly used to provide magnesium ions in reaction systems or as a drying agent for solvents. It is also widely used in preparing buffers and reagents where magnesium is required for enzyme activation. This product is packaged in a 100g bottle, making it ideal for small-scale laboratory experiments. Magnesium sulfate is crucial in analytical procedures like chromatography and protein purification, where maintaining specific ionic conditions is essential for reaction success.
Catalog No. 50-006-61483 Supplier Alkali Scientific Supplier No. MAG003CP
Magnesium Sulfate Heptahydrate (MAG003 CP) is a laboratory-grade salt used in a variety of chemical and biochemical applications. This compound is a crystalline form of magnesium sulfate, commonly used to provide magnesium ions in reaction systems or as a drying agent for solvents. It is also widely used in preparing buffers and reagents where magnesium is required for enzyme activation. This product is packaged in a 100g bottle, making it ideal for small-scale laboratory experiments. Magnesium sulfate is crucial in analytical procedures like chromatography and protein purification, where maintaining specific ionic conditions is essential for reaction success.