Learn More
Alkali Scientific Magnesium Sulfate Heptahydrate, 5 Kg
SDP
Catalog No. 5000661115
Encompass_Preferred
Magnesium Sulfate Heptahydrate (MAG004 CP) is a versatile reagent used in laboratory settings for a variety of applications. This product is supplied in a 5 kg container, ideal for laboratories that require larger quantities for ongoing research. Magnesium sulfate is essential in chemical synthesis, biochemical applications, and as a dehydrating agent. It is commonly used in the preparation of buffers, electrophoresis gels, and as a catalyst in organic reactions. Additionally, magnesium sulfate serves as a stabilizing agent for enzymes and is used in water treatment processes. Its broad utility across multiple disciplines makes it a staple in research and development labs.
Catalog No. 50-006-61115 Supplier Alkali Scientific Supplier No. MAG004CP
Magnesium Sulfate Heptahydrate (MAG004 CP) is a versatile reagent used in laboratory settings for a variety of applications. This product is supplied in a 5 kg container, ideal for laboratories that require larger quantities for ongoing research. Magnesium sulfate is essential in chemical synthesis, biochemical applications, and as a dehydrating agent. It is commonly used in the preparation of buffers, electrophoresis gels, and as a catalyst in organic reactions. Additionally, magnesium sulfate serves as a stabilizing agent for enzymes and is used in water treatment processes. Its broad utility across multiple disciplines makes it a staple in research and development labs.