Learn More
Magnesium Sulfate, Heptahydrate, 98.0 to 102.0%, ACS, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Color | White |
| Physical Form | Crystalline Solid |
| Quantity | 500 g |
| Assay Percent Range | 98.0 to 102.0% |
| Linear Formula | MgSO4 · 7H2O |
| Formula Weight | 246.48 |
| Grade | ACS/EP/Reagent |
| Density | 1.68g/cm3 (20°C); Bulk Density: 900 kg/m3 |
| Chemical Name or Material | Magnesium Sulfate, Heptahydrate, EMSURE™, For Analysis |
Safety and Handling
| Recommended Storage | Store without restrictions |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.