Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate, Heptahydrate, EP, BP, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604135 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
20 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-135 500 g Poly Bottle
18-604-136 2.5 kg Poly Bottle
18-604-137 12 kg Poly Pail
18-604-138 20 kg Fiber Drum
18-604-139 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-135 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA135500GM
Add to Cart
Add to Cart

MA135, 10034-99-8, MgSO4·7H2O

Spectrum™ Chemical Magnesium Sulfate, Heptahydrate, USP, EP, BP is used as an saline laxative. All Spectrum Chemical USP, EP, BP products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

pH 5.0-9.2
CAS Max % 100%
Quantity 500 g
Percent Purity 99 to 100.5%
Grade Multi-Compendial
Loss on Ignition 48.0-52%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.