Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate Heptahydrate, Purified (Epsom Salts), Reagents

Catalog No. 01337183
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-183 50 kg
01-337-186 12 kg
01-337-185 2.5 kg
01-337-184 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-183 Supplier Reagents Holdings Llc Supplier No. C224000050F4
Only null left
Add to Cart
Add to Cart

Magnesium Sulfate Heptahydrate, Purified (Epsom Salts), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Magnesium Sulfate Heptahydrate, Purified (Epsom Salts)
Boiling Point 200°C
CAS 10034-99-8
Color Colorless/White
Physical Form Solid
Quantity 50 kg
Molecular Formula MgSO4, 7H2O
Molecular Weight (g/mol) 246.4746
Grade Pure
Packaging Fiber Drum
Specific Gravity 1.67
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.