Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate Heptahydrate (USP/FCC), Fisher Chemical

Catalog No. M673 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
M67-3 Glass Bottle; 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M67-3

missing translation for 'validMainframeMsg'

Specifications

Melting Point 770°C
Color White
CAS 10034-99-8
pH 5 to 8.2
Physical Form Solid
Quantity 3 kg
Molecular Formula H14MgO11S
MDL Number MFCD00149785
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium(2+) heptahydrate sulfate
Molecular Weight (g/mol) 246.47
PubChem CID 24843
ChEBI CHEBI:31795
Formula Weight 246.48
Percent Purity ≥99.5% (FCC)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (FCC/USP), Pass Test (USP)
Loss on Ignition 40.0 to 52.0% (FCC), 40.0 to 52.0% (FCC/USP), 40.0 to 52.0% (USP)
Packaging Glass Bottle
Vapor Pressure <0.1 mmHg at 20°C
Density 1.678g/cm³
Chemical Name or Material Magnesium Sulfate Heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.