Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate hydrate, Puriss., meets analytical spec. of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance), Solstice

Catalog No. 60044777 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-777 6 x 2.5 kg
60-019-71 1 kg
60-019-72 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-777 Supplier Solstice Supplier No. 131436X2.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of DAC, dried, 99.0-101.0% MgSO4 basis (in dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Quantity 6 x 2.5 kg
Linear Formula MgSO4 · aq
UN Number NONH for all modes of transport
IUPAC Name magnesium;sulfate
Formula Weight 120.37 (anhydrous basis)
Percent Purity 99.0 to 101.0% (MgSO4 basis (in dried substance))
Grade puriss.
Chemical Name or Material Magnesium Sulfate Hydrate, Meets analytical specification of DAC
EINECSNumber 231-298-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.