Learn More
New England Biolabs, Inc. Magnesium Sulfate (MgSO4) Solution – 6 ml (4 x 1.5 ml)
SDP
Catalog No. 504281
Encompass
100 mM MgSO4 for use in conjunction with polymerase reaction buffers.
Catalog No. 50-428-1 Supplier New England Biolabs, Inc. Supplier No. B1003S
100 mM MgSO4 for use in conjunction with polymerase reaction buffers.