Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate, Reagent grade, Solstice

Catalog No. 6002067 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-020-67 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-020-67 Supplier Solstice Supplier No. 2080942.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, anhydrous,≥97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Anhydrous
Quantity 2.5 kg
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
UN Number NONH for all modes of transport
IUPAC Name magnesium;sulfate
Formula Weight 120.37g/mol
Percent Purity ≥97%
Grade Reagent
Chemical Name or Material Magnesium Sulfate
EINECSNumber 231-298-2
RTECSNumber OM4500000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.