Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Magnesium Trifluoromethanesulfonate

Catalog No. 33798625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-798-625G 25 g
33-798-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-798-625G Supplier Sigma Aldrich Supplier No. 33798625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Magnesium Trifluoromethanesulfonate

Specifications

CAS 60871-83-2
Melting Point ≥300°C
Molecular Formula C2F6MgO6S2
Linear Formula (CF3SO3)2Mg
MDL Number MFCD00042601
Quantity 25 g
Synonym Magnesium triflate; Trifluoromethanesulfonic acid magnesium salt
Molecular Weight (g/mol) 322.44 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.