Learn More
Strem, An Ascensus Company CAS# 7439-95-4. 100g. Magnesium turnings for Grignards (99.8%). MFCD00085308
SDP
Catalog No. 5090113983
Encompass
CAS# 7439-95-4. 100g. Magnesium turnings for Grignards (99.8%). MFCD00085308. Molecular Weight: 24.32. Molecular Formula: Mg. Color/form: turnings Strem# 93-1286. http://www.strem.com/catalog/v/93-1286/
Catalog No. 50-901-13983 Supplier Strem, An Ascensus Company Supplier No. 931286100G
CAS# 7439-95-4. 100g. Magnesium turnings for Grignards (99.8%). MFCD00085308. Molecular Weight: 24.32. Molecular Formula: Mg. Color/form: turnings Strem# 93-1286. http://www.strem.com/catalog/v/93-1286/