Learn More
Molecular Devices Molecular Devices Confocal Dual-Disk Box, 60 μm, 50 μm
Catalog No. 5000014417
Encompass_Preferred
Optional dual-disk confocal assembly providing two complementary disk geometries for flexible screening workflows. The unit is designed for rapid swapping and reproducible confocal imaging in high-throughput assays.
  • Dual disk configuration with 60 μm pinhole and 50 μm slit
  • Optimized for basic and high-throughput confocal screening
  • Sealed disk box for stable optical performance
  • Swappable assembly for quick configuration changes
  • Improves assay throughput and reproducibility
Catalog No. 50-000-14417 Supplier Molecular Devices Supplier No. 5074718
Optional dual-disk confocal assembly providing two complementary disk geometries for flexible screening workflows. The unit is designed for rapid swapping and reproducible confocal imaging in high-throughput assays.
  • Dual disk configuration with 60 μm pinhole and 50 μm slit
  • Optimized for basic and high-throughput confocal screening
  • Sealed disk box for stable optical performance
  • Swappable assembly for quick configuration changes
  • Improves assay throughput and reproducibility