Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Magnet for M611 Printer

Catalog No. 17004577 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-004-577 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-004-577 Supplier Brady™ Supplier No. M611MAGNET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use your M611 Label Printer just about anywhere. This magnet attaches the M611 printer to panels, I-beams and other magnetic-receptive surfaces during use or storage.

  • Get hands-free operation by affixing to panel door or metal surface
  • Attaches to back of printer with screws to ensure stability
  • Designed for use with the M611 Label Printer

Specifications

Color Black
For Use With (Equipment) BradyPrinter M611
Width (English) 4.38 in.
Width (Metric) 76.2 cm
Adhesive Type Non-adhesive
Autoclavable Not Autoclavable
Sterility Non-sterile
Includes Magnet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.