Learn More
BUCHI Magnet Valve System, 24 V
SDP
Catalog No. 05405057 Shop All BUCHI Products
Catalog No. 05-405-057 Supplier BUCHI Supplier No. 11060706
Add to Cart
Add to Cart
For use with a centralized vacuum source or an unregulated vacuum pump, compatible with Interface I-300/I-300 Pro.
  • Flow valve without flask
Specifications
Type Magnet Valve
For Use With (Application) Vacuum Regulation of Non-BUCHI Vacuum Pumps or House Vacuums
Voltage 24 V