Learn More
Electron Microscopy Sciences Magnetic Bead Separation Rack for 96-Well PCR Tube Plate
SDP
Catalog No. 501928539
Encompass_Preferred
Magnetic bead separation racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays. They offer exceptional quality and value compared to competing brands. Racks fit microplates following ANSI standards. Clear acrylic allows unobstructed view of tube contents. 24 magnets that hold plate while attracting beads to tube walls.
Catalog No. 50-192-8539 Supplier Electron Microscopy Sciences Supplier No. 6860550
Magnetic bead separation racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays. They offer exceptional quality and value compared to competing brands. Racks fit microplates following ANSI standards. Clear acrylic allows unobstructed view of tube contents. 24 magnets that hold plate while attracting beads to tube walls.