Learn More
Electron Microscopy Sciences EMS Magnetic Induction Stirrer Gray/Purple
SDP
Catalog No. 501931179
Encompass_Preferred
Motorless has high-quality rare-earth magnets to ensure a strong magnetic coupling for vicious solutions. The magnetic coil technology works on the inductive principle with alternative current (AC) as its driving force. The generated moving magnetic field drives the spinning of the stir bar in the vessel. With no moving parts the unit is wear-free and maintenance free and heat is not generated after hours of continuous use making this unit ideal for temperature sensitive samples and environments.
The magnetic field driving the stir bar maintains continuous constant speeds for reproducible results. Control mode, speed, and time with one convenient rotating knob. The stirring speed can be adjusted while a program is running. Includes magnetic induction stirrer, 2 stir bars, 1 silicone anti-skid plate cover, and power adapter.
Color: Gray/Purple
Catalog No. 50-193-1179 Supplier Electron Microscopy Sciences Supplier No. 7236510
Motorless has high-quality rare-earth magnets to ensure a strong magnetic coupling for vicious solutions. The magnetic coil technology works on the inductive principle with alternative current (AC) as its driving force. The generated moving magnetic field drives the spinning of the stir bar in the vessel. With no moving parts the unit is wear-free and maintenance free and heat is not generated after hours of continuous use making this unit ideal for temperature sensitive samples and environments.
The magnetic field driving the stir bar maintains continuous constant speeds for reproducible results. Control mode, speed, and time with one convenient rotating knob. The stirring speed can be adjusted while a program is running. Includes magnetic induction stirrer, 2 stir bars, 1 silicone anti-skid plate cover, and power adapter.
Color: Gray/Purple