Learn More
OHAUS Magnetic 50 Microtube Box
Catalog No. 01804370 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-370 Shaker Attachment
1 options
Catalog No. 01-804-370 Supplier OHAUS Supplier No. 83041370
Add to Cart
Add to Cart
Holds up to 50 microtubes.
  • Attaches to magnetic platform for 360° rotation.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment