Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnetic Animal Cells
SCI_ED

Catalog No. S04227
Change view
Click to view available options
Grade:
4+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04227 4+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04227 Supplier Learning Resources, Inc Supplier No. LER6039
Add to Cart
Add to Cart

Giant cell structures printed on raised foam magnets to appear 3-D.

For demonstration or student manipulation.

  • Four-piece animal cell
  • Fourteen 3-D organelles
  • Activity Guide with background information and diagram master
  • Measures 18 in.
  • Coordinates with foam model (S90816)

Order Info

Grades 4+.

Specifications

For Use With (Application) For demonstration or student manipulation, giant cell structures printed on raised foam Magnets to appear 3D
Grade 4+
Includes 4-piece animal cell, 14 dimensional organelles and activity guide with background information
Size Giant
Product Type Cell Model, Magnetic Animal Cell
Grade Level Elementary, Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.