Learn More
Novus Biologicals™ Magnetic Antibody Labeling Kit

Description
- Conjugates to specially treated magnetic particles
- No extensive optimization of the conjugation reaction
- Covalent attachment of antibodies to magnetic particles
- Bead size: 0.5μm
- Minimizes risk of unspecific binding
Applications: Immunoprecipitation experiments
Storage: Store at 4°C
Note: Kits are guaranteed for 6 months from date of receipt
Specifications
Specifications
| Quantity | 200 μg |
| Product Type | Magnetic antibody labeling kit |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.