Learn More
V&P Scientific Inc MAGNETIC PLATE, 96 Well, Standard or Deep Well, Round or Conical Bottom, 24 Round Posts, BioTek Plate Washer Compatible
Catalog No. NC2177211
Encompass
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 24 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 1 Sided Pellet Location, 4.75mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, BioTek Plate Washer Compatible, Non-SLAS Footprint, No Registration Base
Catalog No. NC2177211 Supplier V&P Scientific Inc Supplier No. VP 771BTSF
Add to Cart
Add to Cart
MAGNETIC BEAD SEPARATION PLATE, 96 Well, Standard or Deep Well Plate, Round or Conical Bottom, 24 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 1 Sided Pellet Location, 4.75mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, BioTek Plate Washer Compatible, Non-SLAS Footprint, No Registration Base