Learn More
V&P Scientific Inc MAGNETIC PLATE, 384 Well, Standard or Deep Well, Flat Bottom, 425 Round Posts
Catalog No. NC1476659
Encompass
MAGNETIC BEAD SEPARATION PLATE, 384 Well, Standard or Deep Well Plate, Flat Bottom, 425 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 4 Sided Pellet Location, 3.1mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, SLAS Footprint, Includes Registration Base
Catalog No. NC1476659 Supplier V&P Scientific Inc Supplier No. VP 771G4AAZM1
MAGNETIC BEAD SEPARATION PLATE, 384 Well, Standard or Deep Well Plate, Flat Bottom, 425 Round Posts, 52MGO, NdFeB Magnets, Red Anodized Aluminum Magnet Frame, 4 Sided Pellet Location, 3.1mm Post Diameter, Axial - North Poles Up Magnetic Orientation, Gripper Accessible, SLAS Footprint, Includes Registration Base