Learn More
V&P Scientific Inc MAGNETIC BEAD EXTRACTION REPLICATOR, MagPin- M
Catalog No. NC0839969
Encompass
MAGNETIC BEAD EXTRACTION REPLICATOR, MagPin M, 96 NdFeB Pins, 48MGO, 3.2mm Diameter, 22mm Long, 9mm Center Spacing, includes a Manual Handle with a Cover Plate Ejector and Loading Jig for VP 407AM-N-PCR
Catalog No. NC0839969 Supplier V&P Scientific Inc Supplier No. VP 407AMN1
MAGNETIC BEAD EXTRACTION REPLICATOR, MagPin M, 96 NdFeB Pins, 48MGO, 3.2mm Diameter, 22mm Long, 9mm Center Spacing, includes a Manual Handle with a Cover Plate Ejector and Loading Jig for VP 407AM-N-PCR