Learn More
V&P Scientific Inc MAGNETIC RACK, 8 Conical Microcentrifuge Tubes, 50mL Volume,
Catalog No. NC1871036
Encompass
MAGNETIC BEAD SEPARATION RACK, 8 Conical Microcentrifuge Tubes, 50mL Volume, 1 Sided Bead Pellet Location, High Position, 48MGO, NdFeB Magnet, 36mm Center Spacing, 29mm Hole Diameter, Polycarbonate
Catalog No. NC1871036 Supplier V&P Scientific Inc Supplier No. VP 772F2M2
MAGNETIC BEAD SEPARATION RACK, 8 Conical Microcentrifuge Tubes, 50mL Volume, 1 Sided Bead Pellet Location, High Position, 48MGO, NdFeB Magnet, 36mm Center Spacing, 29mm Hole Diameter, Polycarbonate