Learn More
Electron Microscopy Sciences Magnetic Bead Separation Rack for 50ml Tube
SDP
Catalog No. 501928536
Encompass_Preferred
Magnetic bead separation racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays. They offer exceptional quality and value compared to competing brands. Racks fit microplates following ANSI standards. Clear acrylic allows unobstructed view of tube contents. Independent sleeves. Spring plunger on tube sleeves for easier liquid addition and removal.
Catalog No. 50-192-8536 Supplier Electron Microscopy Sciences Supplier No. 6860535
Magnetic bead separation racks sequester magnetic beads so that affinity ligands (antibodies, streptavidin or proprietary reagents for coupling nucleic acids) or rinse solutions can be added or decanted in assays. They offer exceptional quality and value compared to competing brands. Racks fit microplates following ANSI standards. Clear acrylic allows unobstructed view of tube contents. Independent sleeves. Spring plunger on tube sleeves for easier liquid addition and removal.