Learn More
MOLECULAR DEPOT LLC Magnetic Beads Buffer - 50 mL
Catalog No. 5000013506
Encompass_Preferred
Magnetic Beads Stabilization Buffer consists of a chemiluminescence buffer extensively tested for the stabilization of a wide variety of magnetic beads (with or without conjugates) This buffer contains a proprietary collection of proteins detergents and other chemicals that promote the long-term stability of magnetic beads For in vitro research use only
Catalog No. 50-000-13506 Supplier MOLECULAR DEPOT LLC Supplier No. B2010186
Magnetic Beads Stabilization Buffer consists of a chemiluminescence buffer extensively tested for the stabilization of a wide variety of magnetic beads (with or without conjugates) This buffer contains a proprietary collection of proteins detergents and other chemicals that promote the long-term stability of magnetic beads For in vitro research use only