Learn More
ACROBiosystems Magnetic Beads™ Streptavidin
Catalog No. 502331502
Encompass_Preferred
The streptavidin magnetic beads are uniform size, narrow size distribution with large surface area and unique surface coating, it is easy to capture the biotinylated proteins or other molecules, and the bounded protein have no activity lost, can help us get the best performance and highly reproducible results.
Catalog No. 50-233-1502 Supplier ACROBiosystems Supplier No. SMBB0110MG
The streptavidin magnetic beads are uniform size, narrow size distribution with large surface area and unique surface coating, it is easy to capture the biotinylated proteins or other molecules, and the bounded protein have no activity lost, can help us get the best performance and highly reproducible results.