Learn More
Instech Protective aluminum cap for magnetic rat Vascular Access Buttons (green)
SDP
Catalog No. NC1680779
Encompass
Instech’s protective aluminum cap for all VABR‑series rat Vascular Access Buttons are engineered with precision, snapping securely into place using magnetic attachment for fast, effortless handling. Its anodized green finish not only delivers standout visibility but also supports group housing of catheterized rats—enhancing animal welfare without compromise.
Catalog No. NC1680779 Supplier Instech Supplier No. VABRCG
Instech’s protective aluminum cap for all VABR‑series rat Vascular Access Buttons are engineered with precision, snapping securely into place using magnetic attachment for fast, effortless handling. Its anodized green finish not only delivers standout visibility but also supports group housing of catheterized rats—enhancing animal welfare without compromise.