Learn More
Grainger Magnetic Catch 20.2 mmx39.9 mmx39.9 mm
Catalog No. 502765032
Encompass_Preferred
The 80/20 Magnetic Catch is a 39.9 mm x 39.9 mm magnetic catch with a holding force of 7 lbs. It is made from zinc alloy with a black finish and has countersunk mounting holes. The catch has two parts - a cup and a ball. The cup attaches to one surface with two screws while the ball attaches to the other surface. When the two surfaces meet, the ball clicks into the cup and is held in place by the magnets. This provides a secure latching mechanism. The catch can be used on boxes, cabinets, and other applications needing a strong magnetic latch. It is RoHS compliant.
Catalog No. 50-276-5032 Supplier Grainger Supplier No. 16U345
The 80/20 Magnetic Catch is a 39.9 mm x 39.9 mm magnetic catch with a holding force of 7 lbs. It is made from zinc alloy with a black finish and has countersunk mounting holes. The catch has two parts - a cup and a ball. The cup attaches to one surface with two screws while the ball attaches to the other surface. When the two surfaces meet, the ball clicks into the cup and is held in place by the magnets. This provides a secure latching mechanism. The catch can be used on boxes, cabinets, and other applications needing a strong magnetic latch. It is RoHS compliant.