Learn More
Brady™ Magnetic Confined Space Cover
Catalog No. 17607640 Shop All Brady Corporation Products
Change view
Click to view available options
Diameter (English):
39 in.
Catalog No. Diameter (English)
17-607-640 39 in.
1 options
Catalog No. 17-607-640 Supplier Brady™ Supplier No. 177437
Add to Cart
Add to Cart
Get a compliant sign and barrier in one. Enhance visibility, reinforce compliance and keep unauthorized personnel out of confined spaces with a Confined Space Cover.
Get a compliant sign and barrier in one. Enhance visibility, reinforce compliance and keep unauthorized personnel out of confined spaces with a Magnetic Confined Space Cover.
  • Built-in magnets secure cover to metal surfaces without the need for fasteners or tools.
  • DANGER sign is compliant with OSHA CFR 1910.146(c)(2) and cover helps prevent unauthorized entry for two-pronged safety.
  • Washable, strong material offers long-lasting durability in various industrial and environmental conditions.
  • Mesh construction helps prevent entry while allowing beneficial airflow.
  • Bright colors and reflective elements enhance visibility.
  • Compact and lightweight for easy transport and storage.
Specifications
Certifications/Compliance OSHA 1910.146
Material Polyester
Description Fits standard 39 in. opening
Diameter (English) 39 in.
Mounting Type Magnetic
Shape Round
Languages English
Displayed Text DANGER
Product Type Confined Space Sign
Color Red