Learn More
Gerstel Magnetic Crimp Caps, PFTE, White, 100/Pack
SDP
Catalog No. NC9278732
Encompass
Bi-Metal, Magnetic, red, Septum: Silicone blue, PTFE white. For seal size 20mm. 100/pk.
Catalog No. NC9278732 Supplier Gerstel Supplier No. 09364006300
Bi-Metal, Magnetic, red, Septum: Silicone blue, PTFE white. For seal size 20mm. 100/pk.