Learn More
Gerstel MAGNETIC CRIMP CAPS 100/PK
SDP
Catalog No. NC9876948
Encompass
Magnetic crimp caps with septa (PTFE red/silicone white/PTFE red) for 2 mL vials, Package of 100
Catalog No. NC9876948 Supplier Gerstel Supplier No. 09364000700
Magnetic crimp caps with septa (PTFE red/silicone white/PTFE red) for 2 mL vials, Package of 100