Learn More
Electron Microscopy Sciences Magnetic Disc Storage Box for AFM, Large
SDP
Catalog No. 501930052
Encompass_Preferred
Store AFM/STM discs in a dust-free environment with these storage boxes. Feature a magnet base to hold the magnetic stainless steel AFM discs and magnetic AFM sample holders. They are made of polystyrene with hinged lids. Perfect for storing, archiving and transporting AFM discs. Carrier only - Discs not included.
Size: Large
Catalog No. 50-193-0052 Supplier Electron Microscopy Sciences Supplier No. 7501203
Store AFM/STM discs in a dust-free environment with these storage boxes. Feature a magnet base to hold the magnetic stainless steel AFM discs and magnetic AFM sample holders. They are made of polystyrene with hinged lids. Perfect for storing, archiving and transporting AFM discs. Carrier only - Discs not included.
Size: Large