Learn More
Supertek™ Scientific Magnetic Field Chamber Model
GREENER_CHOICESCI_ED
Catalog No. S82802 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3+
Catalog No. Grade
S82802 3+
1 options
This item is not returnable. View return policy
Catalog No. S82802 Supplier Supertek™ Scientific Supplier No. PH41272
Add to Cart
Add to Cart
This item is not returnable. View return policy
A hands on scientific magnetic field chamber model for learners
  • Chamber has an internal tunnel into which a bar magnet can be inserted to demonstrate magnetic field pattern
  • Other-shaped magnets can be investigated by positioning their pole near one side of chamber
  • Instructions included
Specifications
Material Plastic
Shape Cylindrical
Grade 3+
Product Type Magnet Activity Set
Includes Transparent acrylic chamber with iron fillings, 1 Cylindrical magnet, and 1 instruction sheet