Learn More
GSC Go Science Crazy Magnetic Field Demonstration
SCI_ED
Catalog No. S28120 Shop All GSC Products
Click to view available options
For Use With (Application):
Helps visualize magnetic field
Catalog No. For Use With (Application)
S28120 Helps visualize magnetic field
1 options
Not a toy. Product intended for ages 13 and up. Magnets not included
Catalog No. S28120 Supplier GSC Go Science Crazy Supplier No. MAGFLD
Not a toy. Product intended for ages 13 and up. Magnets not included
Helps students to understand magnetic fields. Iron filings are housed in a plastic case, and when exposed to magnetic fields begin to disperse along the field lines.
  • Iron filings show magnetic field lines
  • Housed in plastic case
  • Show lines with no mess
  • Instructions included
Specifications
Product Type Magnetic Field Demonstration
Includes Instructions
For Use With (Application) Helps visualize magnetic field
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov