Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Magnetic Field Demonstrator
SCI_ED

Catalog No. S139440 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139440 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139440 Supplier Supertek™ Supplier No. PH41271S3
This item is not returnable. View return policy

A quick and clean way to visualize magnetic fields when using this Supertek™ Magnetic Field Demonstrator.

  • The unit is a transparent acrylic case that holds iron filings in a fluid of proper viscosity.
  • The unit can also be placed on an overhead projector for classroom demonstration.
  • Unit dimensions: 8 x 4-1/2 x 3/8 in.

Specifications

Grade 1+
Format Model
Languages English
Subject Physical Science - Magnetism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.