Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Magnetic Force Science Lab Kit (MAGNETS)
SCI_ED

Catalog No. S02743 Shop All Learning Resources, Inc Products
Change view
Click to view available options
Class Size:
8 to 10 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02743 8 to 10 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02743 Supplier Hand2Mind Supplier No. 90740

missing translation for 'validMainframeMsg'

Students learn the important concepts of magnetic force, levitation, and attraction as they race magnetic cars and build a maglev train, a working compass, and more. This kit also introduces students to different magnet-related STEM careers

  • Explore the science behind magnetism.
  • Use real lab tools and materials to perform amazing experiments and experience what it's like to think like a scientist or an engineer.
  • Become an expert in all things magnetic. The fact-filled guide is chock-full of pictures and information needed for hours of experimentation in your own home laboratory
  • STEM at play kits bring science concepts to life through hands-on activities designed to spark interest, inspire confidence, prompt creative critical-thinking skills while fostering collaboration between children and adults.
  • STEM at play kits provide materials to support scholastic goals while keeping at-home learning fun.
  • 9 Experiments included
    • Work and learn through purposeful science-based play
    • Explore scientific principles
    • Float magnets without touching them
    • Use magnetic levitation to move a train
    • Flip the field on your own iron filings
    • Create a compass ...and more!

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 8 to 10 Students
Product Type Magnetic Force Lab Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.