Learn More
Chemglass Life Sciences Magnetic Glove and Tissue Dispenser Holder
SDP
Catalog No. 5000980655
Encompass
Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, and better abrasion resistance than chloroprene. Better puncture resistance compared to latex or vinyl gloves. Finger-textured finish for improved gripping action. Palm thickness is 4. 5 mils and finger thickness is 5 mils, and standard-length glove is 9. 4" long. Powder-free glove leaves no residue, reducing the risk of contamination compared to a powdered glove. The Bold nitrile, powder-free, disposable glove has better puncture resistance compared to latex or vinyl gloves. Finger-textured finish for improved gripping action. This glove has a palm thickness of 4. 5 mils and a finger thickness of 5 mils, and is 9. 4" long from the tip of the middle finger to the edge of the beaded wrist cuff. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, better abrasion resistance than chloroprene, and reduces the likelihood of allergic reaction. This powder-f
Catalog No. 50-009-80655 Supplier Chemglass Life Sciences Supplier No. CG1790H01
Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, and better abrasion resistance than chloroprene. Better puncture resistance compared to latex or vinyl gloves. Finger-textured finish for improved gripping action. Palm thickness is 4. 5 mils and finger thickness is 5 mils, and standard-length glove is 9. 4" long. Powder-free glove leaves no residue, reducing the risk of contamination compared to a powdered glove. The Bold nitrile, powder-free, disposable glove has better puncture resistance compared to latex or vinyl gloves. Finger-textured finish for improved gripping action. This glove has a palm thickness of 4. 5 mils and a finger thickness of 5 mils, and is 9. 4" long from the tip of the middle finger to the edge of the beaded wrist cuff. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, better abrasion resistance than chloroprene, and reduces the likelihood of allergic reaction. This powder-f