Learn More
World Precision Instrument Magnetic Holding Device - Round base-503568
SDP
Catalog No. 502539470
Encompass
An ideal accessory for optical tables and vibration-free platform. Reduces experimental set-up time by allowing free positioning and instant clamp down of optical components. Switchable ON/OFF magnetic circuit permits fine adjustment and precise positioning.
Catalog No. 50-253-9470 Supplier World Precision Instrument Supplier No. 503568
An ideal accessory for optical tables and vibration-free platform. Reduces experimental set-up time by allowing free positioning and instant clamp down of optical components. Switchable ON/OFF magnetic circuit permits fine adjustment and precise positioning.