Learn More
Bellco Glass, Inc. Magnetic Mixer, Double Ended, 8mm Diameter, 55mm Long, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800423
Encompass_Preferred
Magnetic Mixers are used to mix components in order to obtain a homogeneous liquid mixtures. Our mixers can be used in a variety of applications including but not limited to chemistry and biology experiments, buffer solutions, and bacterial growth media. magnetic mixers are ideal for mixing over a long period of time. Since our mixers are just magnet bars, they can easily be cleaned and the risks of contaminating the solution can be minimized. Diameter: 8 mm, Length: 55 mm
Catalog No. 50-255-0339