Learn More
McMaster-Carr Magnetic Brackets
Catalog No. NC2392913
Encompass
The McMaster-Carr Magnetic Brackets are made of steel with a zinc plating to resist corrosion. They have extremely strong rare earth magnets on the back to adhere to ferrous metal surfaces. The brackets can support up to 50 lbs loads on vertical surfaces when used in pairs. They are 1" wide x 2" high x 1/2" deep. The magnets measure 1/2" dia x 1/4" thick. They allow you to mount things on metal without drilling holes. Typical applications include mounting signs, hooks, and more on surfaces like toolboxes, shelving, cabinets, and equipment.
Catalog No. NC2392913 Supplier McMaster-Carr Supplier No. 2330N21
The McMaster-Carr Magnetic Brackets are made of steel with a zinc plating to resist corrosion. They have extremely strong rare earth magnets on the back to adhere to ferrous metal surfaces. The brackets can support up to 50 lbs loads on vertical surfaces when used in pairs. They are 1" wide x 2" high x 1/2" deep. The magnets measure 1/2" dia x 1/4" thick. They allow you to mount things on metal without drilling holes. Typical applications include mounting signs, hooks, and more on surfaces like toolboxes, shelving, cabinets, and equipment.