Learn More
Revvity Health Sciences Inc Magnetic Quick Connect
Catalog No. 502016498
Encompass
Magnetic Quick Connect is the conduit for connection of multiple anesthesia delivery accessories to the IVIS Lumina S5 and IVIS Lumina X5 imaging systems including the SRM-5 (Small Rodent Manifold) and the SMT-5 (Smart Mouse Tray) (both sold separately). Simply slide accessory onto quick connect ports and magnets with pull accessory into place and engage anesthesia. Facilitates high throughput imaging features via easy connection/disconnection of anesthesia accessories.
Catalog No. 50-201-6498 Supplier Revvity Health Sciences Inc Supplier No. CLS148876
Add to Cart
Add to Cart
Request a Quote
Magnetic Quick Connect is the conduit for connection of multiple anesthesia delivery accessories to the IVIS Lumina S5 and IVIS Lumina X5 imaging systems including the SRM-5 (Small Rodent Manifold) and the SMT-5 (Smart Mouse Tray) (both sold separately). Simply slide accessory onto quick connect ports and magnets with pull accessory into place and engage anesthesia. Facilitates high throughput imaging features via easy connection/disconnection of anesthesia accessories.