Learn More
Grainger Ullman Devices Magnetic Pick-Up Tool, Telescoping, 1.5 lb. Capacity, 1 ea
Catalog No. 502780015
Encompass_Preferred
This pocket-sized magnetic pick-up tool is designed for the rapid and accurate retrieval of small metal objects from restricted or inaccessible areas. Featuring a telescoping shaft and an adjustable, rotating magnetic head, this tool provides precise maneuverability for various maintenance and inspection tasks.
  • Adjustable rotating magnetic head
  • Magnetic lift capacity of 1.5 lb.
  • Telescoping shaft extends from 16.2 cm (6-3/8 in.) to 48.3 cm (19 in.)
  • Constructed from durable stainless steel
  • Integrated pocket clip for secure transport and accessibility
Catalog No. 50-278-0015 Supplier Grainger Supplier No. 19LZ14
This pocket-sized magnetic pick-up tool is designed for the rapid and accurate retrieval of small metal objects from restricted or inaccessible areas. Featuring a telescoping shaft and an adjustable, rotating magnetic head, this tool provides precise maneuverability for various maintenance and inspection tasks.
  • Adjustable rotating magnetic head
  • Magnetic lift capacity of 1.5 lb.
  • Telescoping shaft extends from 16.2 cm (6-3/8 in.) to 48.3 cm (19 in.)
  • Constructed from durable stainless steel
  • Integrated pocket clip for secure transport and accessibility