Learn More
Grainger Magnetic Retrieval Tool 6 5/8 -33 1/4
Catalog No. 502765229
Encompass_Preferred
The Ullman Magnetic Retrieval Tool is a telescoping magnetic pickup tool designed to retrieve metal objects from hard-to-reach places. It extends from 15 to 34 inches and has a powerful rare earth magnet on the end that can hold up to 5 pounds. The aluminum body is lightweight yet durable. This versatile tool can be used to pick up nuts, bolts, screws, nails, tools, car parts and more in tight spaces. It's great for automotive work, construction sites, workshops and around the home. The rubber grip provides comfort and control.
Catalog No. 50-276-5229 Supplier Grainger Supplier No. 16W181
The Ullman Magnetic Retrieval Tool is a telescoping magnetic pickup tool designed to retrieve metal objects from hard-to-reach places. It extends from 15 to 34 inches and has a powerful rare earth magnet on the end that can hold up to 5 pounds. The aluminum body is lightweight yet durable. This versatile tool can be used to pick up nuts, bolts, screws, nails, tools, car parts and more in tight spaces. It's great for automotive work, construction sites, workshops and around the home. The rubber grip provides comfort and control.