Learn More
Grainger Ullman Magnetic Retrieval Tool, Neodymium, Telescoping, 1 in. Jaw, 1 ea.
Catalog No. 502766192
Encompass_Preferred
This telescoping magnetic retrieval tool is designed for efficient recovery of metal parts and debris in restricted or hard-to-reach areas. Featuring a powerful neodymium magnet and a stainless steel construction, it provides reliable performance in industrial and workshop environments. The tool includes a protective cap to focus magnetic force, minimizing unwanted side attraction.
  • Telescoping handle extends from 7-1/4 in. to 33-3/4 in.
  • Neodymium-iron-boron magnet provides 5 lb. lift capacity.
  • Magnetic power increased by 45% or more using included shield.
  • Cushioned, non-slip grip prevents drops in oily or wet conditions.
  • Convenient pocket clip for easy transport.
Catalog No. 50-276-6192 Supplier Grainger Supplier No. 16W182
This telescoping magnetic retrieval tool is designed for efficient recovery of metal parts and debris in restricted or hard-to-reach areas. Featuring a powerful neodymium magnet and a stainless steel construction, it provides reliable performance in industrial and workshop environments. The tool includes a protective cap to focus magnetic force, minimizing unwanted side attraction.
  • Telescoping handle extends from 7-1/4 in. to 33-3/4 in.
  • Neodymium-iron-boron magnet provides 5 lb. lift capacity.
  • Magnetic power increased by 45% or more using included shield.
  • Cushioned, non-slip grip prevents drops in oily or wet conditions.
  • Convenient pocket clip for easy transport.