Learn More
Grainger Ullman Devices Magnetic Pick-Up Tool, Telescoping, 16 lb Capacity, 1 ea
Catalog No. 502766421
Encompass_Preferred
The MegaMag® telescoping magnetic pick-up tool is designed for the precise retrieval of lost metal objects in hard-to-reach areas. Featuring a specialized goblet-shaped head, this tool effectively shields the sides of the magnet to prevent unintended attachment to surrounding metal surfaces. The device is constructed with a durable stainless steel handle and includes a comfortable cushion grip to ensure stability and prevent slipping during use in wet or oily conditions.
  • Telescopes from 8-1/4 in. to 30-1/4 in. for extended reach.
  • Supports a magnetic lifting capacity of up to 16 lbs.
  • Features a goblet-shaped neodymium magnet head that shields sides to prevent unwanted attachment.
  • Equipped with a non-slip, textured cushion grip for secure handling.
  • Stainless steel construction ensures durability.
Catalog No. 50-276-6421 Supplier Grainger Supplier No. 16W179
The MegaMag® telescoping magnetic pick-up tool is designed for the precise retrieval of lost metal objects in hard-to-reach areas. Featuring a specialized goblet-shaped head, this tool effectively shields the sides of the magnet to prevent unintended attachment to surrounding metal surfaces. The device is constructed with a durable stainless steel handle and includes a comfortable cushion grip to ensure stability and prevent slipping during use in wet or oily conditions.
  • Telescopes from 8-1/4 in. to 30-1/4 in. for extended reach.
  • Supports a magnetic lifting capacity of up to 16 lbs.
  • Features a goblet-shaped neodymium magnet head that shields sides to prevent unwanted attachment.
  • Equipped with a non-slip, textured cushion grip for secure handling.
  • Stainless steel construction ensures durability.