Learn More
Grainger Magnetic Retrieval Tool 8 1/4 -30 1/4
Catalog No. 502766416
Encompass_Preferred
The Ullman Magnetic Retrieval Tool is a telescoping magnetic pick-up tool designed to retrieve metal objects from hard-to-reach places. It features a strong magnet on the end of an aluminum telescoping pole that extends from 15 to 48 inches. The head rotates 360 degrees for picking up objects at any angle. It can lift up to 5 pounds and collapses to 15 inches for easy storage. The knurled aluminum handle provides a comfortable grip. This versatile tool is great for retrieving dropped nuts, bolts, tools, and other metal objects from tight spaces.
Catalog No. 50-276-6416 Supplier Grainger Supplier No. 16W183
The Ullman Magnetic Retrieval Tool is a telescoping magnetic pick-up tool designed to retrieve metal objects from hard-to-reach places. It features a strong magnet on the end of an aluminum telescoping pole that extends from 15 to 48 inches. The head rotates 360 degrees for picking up objects at any angle. It can lift up to 5 pounds and collapses to 15 inches for easy storage. The knurled aluminum handle provides a comfortable grip. This versatile tool is great for retrieving dropped nuts, bolts, tools, and other metal objects from tight spaces.