Learn More
MSC PEC Tools Rigid Magnetic Rule, 24 in., 5R Graduation, Satin Chrome Finish, 1 ea.
Catalog No. 5000512820
Encompass
These rigid magnetic rules are precision tools designed for hands-free measurement in metalworking applications. Constructed from steel with a durable satin chrome finish, these rules feature high-powered magnets inset at 6 in. intervals to allow for secure attachment to machine surfaces or toolboxes.
  • Features high-powered magnets every 6 in. for hands-free operation
  • Constructed from steel for durability
  • Satin chrome finish for readability
  • Includes 5R graduations
  • Measures 24 in. in length and 1.25 in. in width
  • Originates from USA
Catalog No. 50-005-12820 Supplier MSC Supplier No. 07154545
These rigid magnetic rules are precision tools designed for hands-free measurement in metalworking applications. Constructed from steel with a durable satin chrome finish, these rules feature high-powered magnets inset at 6 in. intervals to allow for secure attachment to machine surfaces or toolboxes.
  • Features high-powered magnets every 6 in. for hands-free operation
  • Constructed from steel for durability
  • Satin chrome finish for readability
  • Includes 5R graduations
  • Measures 24 in. in length and 1.25 in. in width
  • Originates from USA